Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Disposable IR Cards, 5x10cm, KBr window, 19mm aperture

Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.

$118.30 - $298.00

Specifications

Description Disposable IR Cards
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA43935KJ
View Documents
Alfa Aesar™
43935KJ
5 Pk. Each for $118.30
Order in Bulk

Request bulk or custom formats

Add to cart
 
AA43935KM
View Documents
Alfa Aesar™
43935KM
25 Pk. Each for $298.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

  • Size: 5 x 10cm
  • KBr window
  • 19mm aperture
Specifications

Specifications

Disposable IR Cards
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit