Alfa Aesar™ Disposable IR Cards, 5x10cm, KCl window, 19mm aperture

Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.

Manufacturer: Alfa Aesar™ 43932KJ

 View more versions of this product

Catalog No. AA43932KJ

  Check Availability
Add to Cart

    Specifications

    Quantity 5 Pk.
    Description Disposable IR Cards

    • Size: 5 x 10cm
    • KCl window
    • 19mm aperture

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit