Learn More
GE Healthcare Disposable PD-10 Columns: Pk of 50 Empty Columns Promotion
PD-10 Columns are economical, plastic columns for gravity flow purification
Manufacturer:  GE Healthcare 29334693
Catalog No. 45-000-067PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Economical and versatile columns for gravity flow applications
  • Empty PD-10 columns can be packed with a wide range of bulk media
  • Easy to use
  • Use PD-10 Columns with LabMate Buffer Reservoir for convenient equilibration, sample application, and wash
  • Empty Disposable PD-10 Columns are economical, plastic columns for single-use applications using gravity flow
  • The columns hold up to approx. 13 ml of chromatography media; the frits can be used as an upper bed support that prevents the medium from running dry under gravity flow
  • Comes with caps
Specifications
Specifications
50 Pack
0.59 in.
1.5cm
Empty Columns; comes with caps
Designed for ease of use and equilibration