Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Disposable PD-10 Desalting Columns: PROMOTION

Use for desalting and buffer exchange, as well as removal of low-molecular weight compounds. GE Healthcare Life Sciences™ Disposable PD-10 Desalting Columns provide fast, high-recovery.

Manufacturer:  GE Healthcare 29334696

Catalog No. 45-000-148PM

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Packed with Sephadex™ G-25 medium
  • Designed for ease of use and equilibration
  • Prepare samples in range of 1.0 to 2.5mL by gravity flow or 1.75 to 2.5mL by centrifugation
  • Desalt multiple samples in parallel with desalting capacity >90%
  • Sample recovery in range of 70 to >95%
  • Includes adapters for centrifugation
Specifications

Specifications

30 Pack
>50μm (Dry), 38 to 235μm (wet)
Designed for ease of use and equilibration
4° to 30°C
Desalting column packed with Sephadex G-25 medium
0.59 in.
1.5cm
8.3mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit