Learn More
Fisherbrand™ Disposable Plain-End Flint Glass Tubes with Color-Coded Label Area
Eliminate the problem of lost or peeling labels with these disposable, color-coded tubes.
$139.21 - $263.04
Specifications
Closure | KIM-KAP |
---|---|
Material | Flint Glass |
Shape | Round Bottom |
Disposable | Yes |
Type | Disposable Culture Tube |
Catalog Number | Color | Diameter (Metric) Outer | Length (Metric) | Capacity (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Color | Diameter (Metric) Outer | Length (Metric) | Capacity (Metric) | Price | Quantity & Availability | |||
14-957-10A
|
White | 10 mm | 75 mm | 4 mL |
Case of 1000 for $139.21
|
|
14-957-10B
|
Yellow | 10 mm | 75 mm | 4 mL |
Case of 1000 for $141.35
|
|
14-957-10C
|
Green | 10 mm | 75 mm | 4 mL |
Case of 1000 for $142.60
|
|
14-957-10D
|
Orange | 10 mm | 75 mm | 4 mL |
Case of 1000 for $142.60
|
|
14-957-10E
|
Blue | 10 mm | 75 mm | 4 mL |
Case of 1000 for $142.60
|
|
14-957-12A
![]() |
White | 12 mm | 75 mm | 6 mL |
Case of 1000 for $141.36
|
|
14-957-12B
![]() |
Yellow | 12 mm | 75 mm | 6 mL |
Case of 1000 for $142.61
|
|
14-957-12C
|
Green | 12 mm | 75 mm | 6 mL |
Case of 1000 for $142.61
|
|
14-957-12D
|
Orange | 12 mm | 75 mm | 6 mL |
Case of 1000 for $142.61
|
|
14-957-12E
|
Blue | 12 mm | 75 mm | 6 mL |
Case of 1000 for $142.61
|
|
14-957-14A
|
White | 13 mm | 100 mm | 10 mL |
Case of 1000 for $169.53
|
![]() |
14-957-14B
|
Yellow | 13 mm | 100 mm | 10 mL |
Case of 1000 for $169.53
|
|
14-957-14D
|
Orange | 13 mm | 100 mm | 10 mL |
Case of 1000 for $169.53
|
|
14-957-14E
![]() |
Blue | 13 mm | 100 mm | 10 mL |
Case of 1000 for $169.53
|
|
14-957-16A
|
White | 16 mm | 100 mm | 15 mL |
Case of 1000 for $210.11
|
|
14-957-16B
|
Yellow | 16 mm | 100 mm | 15 mL |
Case of 1000 for $210.11
|
|
14-957-16C
|
Green | 16 mm | 100 mm | 15 mL |
Case of 1000 for $210.11
|
|
14-957-16D
|
Orange | 16 mm | 100 mm | 15 mL |
Case of 1000 for $210.11
|
|
14-957-16E
|
Blue | 16 mm | 100 mm | 15 mL |
Case of 1000 for $210.11
|
|
14-957-18D
|
Orange | 16 mm | 125 mm | 19 mL |
Case of 1000 for $252.75
|
|
14-957-18E
![]() |
Blue | 16 mm | 125 mm | 19 mL |
Case of 1000 for $252.75
|
|
14-957-18A
|
White | 16 mm | 125 mm | 19 ml |
Case of 1000 for $263.04
|
|
Description
Eliminate the problem of lost or peeling labels with these disposable, color-coded tubes.
- Fused-in, textured, color patches accept any type of ink or pencil
- Expose to heat, steam, hours in a water bath, without affecting label
- Marking area is large enough for all necessary information—1.37 in. (for 10 and 12mm O.D. tubes) or 2 in. (for 13 and 16mm O.D. tubes) long labeling area in five pastel colors
- Round bottom
- Flint or borosilicate glass
Specifications
KIM-KAP | |
Round Bottom | |
Disposable Culture Tube |
Flint Glass | |
Yes |