Learn More
Fisherbrand™ Disposable Plain-End Glass Tubes with Color-Coded Label Area
SDP
Eliminate the problem of lost or peeling labels with these disposable, color-coded tubes.
$165.00 - $262.00
Specifications
Closure Type KIM-KAP
Product Type Disposable Culture Tube
Material Borosilicate Glass
Disposable Yes
For Use With End problem of lost or peeling labels
View More Specs
Products 19
Catalog Number Color Diameter (Metric) Length (Metric) Capacity (Metric) Price Quantity & Availability  
Catalog Number Color Diameter (Metric) Length (Metric) Capacity (Metric) Price Quantity & Availability  
14-957-20A
SDP
View Documents
White 10 mm 75 mm 4 mL
Case of 1000 for $165.00
Add to Cart
14-957-20B
SDP
View Documents
Yellow 10 mm 75 mm 4 mL
Case of 1000 for $165.00
Add to Cart
14-957-20C
SDP
View Documents
Green 10 mm 75 mm 4.0 mL
Case of 1000 for $165.00
Add to Cart
14-957-20E
SDP
View Documents
Blue 10 mm 75 mm 4 mL
Case of 1000 for $165.00
Add to Cart
14-957-20D
SDP
View Documents
Orange 10 mm 75 mm 4 mL
Case of 1000 for $165.00
Add to Cart
14-957-22A
SDP
View Documents
White 12 mm 75 mm 6 mL
Case of 1000 for $180.00
Add to Cart
14-957-22B
SDP
View Documents
Yellow 12 mm 75 mm 6 mL
Case of 1000 for $180.00
Add to Cart
14-957-22C
SDP
View Documents
Green 12 mm 75 mm 6 mL
Case of 1000 for $180.00
Add to Cart
14-957-22E
SDP
View Documents
Blue 12 mm 75 mm 6 mL
Case of 1000 for $180.00
Edge
Add to Cart
14-957-24A
SDP
Accessories View Documents
White 13 mm 100 mm 10 mL
Case of 1000 for $212.00
Add to Cart
14-957-22D
SDP
View Documents
Orange 12 mm 75 mm 6 mL
Case of 1000 for $180.00
Add to Cart
14-957-24B
SDP
Accessories View Documents
Yellow 13 mm 100 mm 10 mL
Case of 1000 for $212.00
Add to Cart
14-957-24C
SDP
Accessories View Documents
Green 13 mm 100 mm 10 mL
Case of 1000 for $212.00
Add to Cart
14-957-24E
SDP
Accessories View Documents
Blue 13 mm 100 mm 10 mL
Case of 1000 for $212.00
Edge
Add to Cart
14-957-24D
SDP
Accessories View Documents
Orange 13 mm 100 mm 10 mL
Case of 1000 for $212.00
Add to Cart
14-957-26A
SDP
Accessories View Documents
White 16 mm 100 mm 15 mL
Case of 1000 for $262.00
Edge
Add to Cart
14-957-26B
SDP
Accessories View Documents
Yellow 16 mm 100 mm 15 mL
Case of 1000 for $262.00
Add to Cart
14-957-26E
SDP
Accessories View Documents
Blue 16 mm 100 mm 15 mL
Case of 1000 for $262.00
Add to Cart
14-957-26C
SDP
Accessories View Documents
Green 16 mm 100 mm 15 mL
Case of 1000 for $262.00
Add to Cart
Description
Description
Eliminate the problem of lost or peeling labels with these disposable, color-coded tubes.
  • Fused-in, textured, color patches accept any type of ink or pencil
  • Expose to heat, steam, hours in a water bath, without affecting label
  • Marking area is large enough for all necessary information—1.37 in. (for 10 and 12mm O.D. tubes) or 2 in. (for 13 and 16mm O.D. tubes) long labeling area in five pastel colors
  • Round bottom
Specifications
Specifications
KIM-KAP
Borosilicate Glass
End problem of lost or peeling labels
Disposable Culture Tube
Yes
Non-sterile