Learn More
Disposable Polyethylene Food Service Gloves
Resistant to most chemicals, reagents and acids.
Specifications
Material | Polyethylene |
---|---|
Glove Cuff Style | XL |
Powdered | Powder-free |
Thickness (Metric) | 1 mm |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
S47328
![]() |
AEP Industries
2GM1F |
Small |
Pack of 100 for $4.70
Case of 10 PK for $47.00 |
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
S47330
![]() |
AEP Industries
2GM2F |
Medium |
Pack of 100 for $4.70
Case of 10 PK for $47.00 |
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
S47332
![]() |
AEP Industries
2GM3F |
Large |
Pack of 100 for $4.70
Case of 10 PK for $47.00 |
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
Description
For non-precision food preparation and service applications
- Made of polyethylene-blended film
- Extra-long cuffs
- Medium weight (1mil thickness)
- Tough and durable, premium-weight, soft embossed glove
- Easy-on and off for jobs requiring frequent changes
- Textured for enhanced grip and hand comfort
- Economical, lightweight
Specifications
Polyethylene | |
XL | |
Powder-free | |
1 mm |