Learn More
Scientific Industries DISRUPTOR BEADS 0.5MM 8 OZ.
Supplier:  Scientific Industries SIBG05
Encompass_Preferred
Glass beads are designed for use with the Disruptor Genie or TurboMix attachment (used with Vortex-Genie 2 or Vortex-Genie 2T) for optimum disruption of bacteria, yeast, fungi and other cells. .5 mm ideal for yeast,plant tissue.
Catalog No. 50-728-198
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.