Learn More
Dissecting Needles, Probes, and Malls Products for Science Education
Help students master dissecting techniques — choose the right needle and probe for the job.
Manufacturer:  Medco Instruments 0137CH
 View more versions of this product
Catalog No. S17344
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Our selection of dissecting needles and probes includes all high-quality stainless-steel instruments with precision points for precision work.
Teasing Needle with Metal Holder
  • 5cm long needle holder with an adjustable chuck for tightening or replacing the needles
  • Includes one straight needle
  • Replacement needles available separately (Bent — S17340B and Straight — S17340S, not pictured)
Flexible Probe with Eye
  • One end of this 152mm probe is olive-tapered
  • Other end is flat with an eye
Seeker Mall Probe
  • 152mm mall probe fashioned from chrome-plated steel with a hexagonal handle
  • One end is tapered to an angled point
  • Other end is blunt
Probe and Hook
  • Eight-sided handle on probe for easy turning and gripping while dissecting
  • Curved tenaculum end has a sharpened inside edge for cutting
  • Straight, tapered point is also sharp
Huber Probe
  • Double-ended probe and seeker
  • Has angular, tapered probe on one end
  • Other end has straight, tapered probe
Specifications
Specifications
6 in.
15.24cm
Huber Probe
Precision work, help students master dissecting techniques
Stainless Steel