Learn More
Dissecting Needles, Probes, and Malls
Help students master dissecting techniques choose the right needle and probe for the job.
Manufacturer: Medco Instruments 83702
Description
Our selection of dissecting needles and probes includes all high-quality stainless-steel instruments with precision points for precision work.
Teasing Needle with Metal Holder
- 5cm long needle holder with an adjustable chuck for tightening or replacing the needles
- Includes one straight needle
- Replacement needles available separately (Bent S17340B and Straight S17340S, not pictured)
- One end of this 152mm probe is olive-tapered
- Other end is flat with an eye
Seeker Mall Probe
- 152mm mall probe fashioned from chrome-plated steel with a hexagonal handle
- One end is tapered to an angled point
- Other end is blunt
Probe and Hook
- Eight-sided handle on probe for easy turning and gripping while dissecting
- Curved tenaculum end has a sharpened inside edge for cutting
- Straight, tapered point is also sharp
Huber Probe
- Double-ended probe and seeker
- Has angular, tapered probe on one end
- Other end has straight, tapered probe
Specifications
Precision work, help students master dissecting techniques | |
15.24cm | |
Flexible Probe |
6 in. | |
Stainless Steel |