Learn More
Dissection Pans and Pads
SDPSCI_ED
Keep specimens looking fresh between dissection periods.
Specifications
For Use With (Application) Dissecting
View More Specs
Products 7
Catalog Number Mfr. No. Material Product Type Includes Price Quantity & Availability  
Catalog Number Mfr. No. Material Product Type Includes Price Quantity & Availability  
S17406
SDPSCI_ED
View Documents
Nasco
SB14451DI
Vinyl Econo Pan Flex-pad This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.This product is either new or inactive, and cannot be ordered online. N/A
S17408
SDPSCI_ED
View Documents
Nasco
SB14452DIM
Vinyl Flex-pad Flex-pad only (no pan) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.This product is either new or inactive, and cannot be ordered online. N/A
S17471
SDPSCI_ED
View Documents
Nasco
SB15252FIM
HDPE, Vinyl Deluxe Pan Flex-pad and cover This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.This product is either new or inactive, and cannot be ordered online. N/A
S17468
SDPSCI_ED
View Documents
Nasco
SB15251FIM
HDPE, Vinyl Standard Pan Flex-pad and cover This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.This product is either new or inactive, and cannot be ordered online. N/A
S17469
SDPSCI_ED
View Documents
Nasco
SB15233FIM
HDPE, Vinyl Standard Pan Flex-pad This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
S17472
SDPSCI_ED
View Documents
Nasco
SB15236FIM
HDPE, Vinyl Deluxe Pan Flex-pad This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.This product is either new or inactive, and cannot be ordered online. N/A
S17473
SDPSCI_ED
View Documents
Nasco
SB15237DI
HDPE Deluxe Pan Pan only This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.This product is either new or inactive, and cannot be ordered online. N/A
Description
Description
  • High-density polyethylene (HDPE) pans offer tight-sealing lids for durability and easy cleanup
  • Each is available with Flex-Pads — odorless vinyl pads that are machine washable and reusable
  • Flex-Pads replace wax for faster setup and cleanup
Specifications
Specifications
Dissecting