Learn More
LaMotte Dissolved Oxygen Kit
Learn how to conduct oxygen-level tests for plants and animals.
$39.00 - $62.50
Specifications
For Use With (Application) | Water Testing |
---|---|
For Use With (Equipment) | Dissolved oxygen kit |
Includes: Five reagents; makes 50 tests at 10ppm
Catalog Number | Mfr. No. | Product Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Includes | Price | Quantity & Availability | ||||
S45088R
![]() ![]() |
LaMotte
R5860 |
Dissolved Oxygen Kit Reagent Refill | 5 Reagents |
Each for $39.00
|
|
S94979
![]() ![]() |
LaMotte
586001 |
Dissolved Oxygen Kit | 30mL Manganous Sulfate Solution, 30mL Alkaline Potassium Iodide Azide, 30mL Sulfuric Acid, 60mL 0.025N Sodium Thiosulfate, 30mL Starch indicator solution, direct reading titrator, 5-10-12.9-15-20-25mL glass test tube with cap, 60mL water sampling Glass bottle |
Each for $62.50
|
|
Description
- Direct-reading titrator test measures levels of oxygen
- Indicates whether a body of water can sustain healthy aquatic plants and animals
- Low oxygen levels signal contamination from organic wastes
- Measures 0 to 10ppm in 0.2ppm increments
- Azide modification of Winkler method
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Water Testing | |
Dissolved oxygen kit |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.