Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Distemper Virus Mouse anti-Virus, DyLight 680, Clone: 5-4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB1102604FR

Catalog No. NB1102604FR


Add to cart

Description

Description

Distemper Virus Monoclonal specifically detects Distemper Virus in Virus samples. It is validated for ELISA, Sandwich ELISA.
Specifications

Specifications

Distemper Virus
43224
ELISA, Sandwich ELISA
Purified
Tissue cell preparation.
Protein A purified
RUO
Primary
Specific for surface envelope antigen of CDV
ELISA
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG2a
0.2 ml
Store at 4C in the dark.
Monoclonal
Virus
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit