Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disulfiram, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum

D1389, 97-77-8, C10H20N2S4

Spectrum™ Disulfiram, USP is used to treat chronic alcoholism by creating acute sensitivity to alcohol in the body. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Fiber Drum
Ignition Residue 0.001
Quantity 25 kg
Melting Range 69-72°C
CAS 97-77-8
CAS Max % 1
Molecular Formula C10H20N2S4
InChI Key AUZONCFQVSMFAP-UHFFFAOYSA-N
SMILES CCN(CC)C(=S)SSC(=S)N(CC)CC
IUPAC Name N,N-diethyl[(diethylcarbamothioyl)disulfanyl]carbothioamide
Molecular Weight (g/mol) 296.52
Percent Purity 98 to 102%
Show More Show Less