Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dithiobiuret, 97%

Click to view available options
Quantity:
5g
25g
100g
541-53-7
C2H5N3S2
135.203
MFCD00037832
JIRRNZWTWJGJCT-UHFFFAOYSA-N
dithiobiuret,2,4-dithiobiuret,thioimidodicarbonic diamide,biuret, dithio,imidodicarbonodithioic diamide,biuret, 2,4-dithio,imidodicarbonimidothioic diamide,rcra waste number p049,urea, 2-thio-1-thiocarbamoyl,allophanimidic acid, dithio
2758725
carbamothioylthiourea
C(=S)(N)NC(=S)N
541-53-7
C2H5N3S2
135.203
MFCD00037832
JIRRNZWTWJGJCT-UHFFFAOYSA-N
dithiobiuret,2,4-dithiobiuret,thioimidodicarbonic diamide,biuret, dithio,imidodicarbonodithioic diamide,biuret, 2,4-dithio,imidodicarbonimidothioic diamide,rcra waste number p049,urea, 2-thio-1-thiocarbamoyl,allophanimidic acid, dithio
2758725
carbamothioylthiourea
C(=S)(N)NC(=S)N
Odor Odorless
Melting Point ~183°C (decomposition)
Quantity 5g
UN Number UN2811
Beilstein 1756621
Merck Index 14,3371
Formula Weight 135.21
Percent Purity 97%
Chemical Name or Material Dithiobiuret

GHS H Statement
H300
Fatal if swallowed.

P264b-P270-P301+P310-P330-P501c

H300

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 208-784-8

RTECSNumber : EC1575000

TSCA : Yes

Recommended Storage : Ambient temperatures