Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dithioerythritol, 99+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
6892-68-8
C4H10O2S2
154.25
MFCD00063750
VHJLVAABSRFDPM-ZXZARUISSA-N
dithioerythritol,1,4-dithioerythritol,dte,2r,3s-1,4-dimercaptobutane-2,3-diol,erythro-1,4-dimercapto-2,3-butanediol,2,3-butanediol, 1,4-dimercapto-, 2r,3s-rel,1,4-dithioerythritol dte,2r*,3s*-1,4-dimercapto-2,3-butanediol,erythro-2,3-dihydroxy-1,4-butanedithiol,2r,3s-1,4-disulfanylbutane-2,3-diol
439352
CHEBI:17456
(2R,3S)-1,4-bis(sulfanyl)butane-2,3-diol
C(C(C(CS)O)O)S
6892-68-8
C4H10O2S2
154.25
MFCD00063750
VHJLVAABSRFDPM-ZXZARUISSA-N
dithioerythritol,1,4-dithioerythritol,dte,2r,3s-1,4-dimercaptobutane-2,3-diol,erythro-1,4-dimercapto-2,3-butanediol,2,3-butanediol, 1,4-dimercapto-, 2r,3s-rel,1,4-dithioerythritol dte,2r*,3s*-1,4-dimercapto-2,3-butanediol,erythro-2,3-dihydroxy-1,4-butanedithiol,2r,3s-1,4-disulfanylbutane-2,3-diol
439352
CHEBI:17456
(2R,3S)-1,4-bis(sulfanyl)butane-2,3-diol
C(C(C(CS)O)O)S
Infrared Spectrum Authentic
Appearance (1% in water) colorless, clear to very slightly hazy
Color White to Yellow or Beige
Packaging Glass bottle
Melting Point 82°C to 86°C
Quantity 5g
Assay Percent Range 99% min. (Iodimetry)
Linear Formula HSCH2CH(OH)CH(OH)CH2SH
Beilstein 01, III, 2359
Solubility Information (1% in water) Clear to slightly hazy colorless
Formula Weight 154.25
Physical Form Powder
Percent Purity 99+%
Chemical Name or Material Dithioerythritol
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 229-998-8