Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dithiooxamide, 98%, Acros Organics™

Click to view available options
Quantity:
5g
25g
100g
79-40-3
C2H4N2S2
120.188
OAEGRYMCJYIXQT-UHFFFAOYSA-N
dithiooxamide,rubeanic acid,ethanebis thioamide,oxamide, dithio,dithioxamide,rubeane,rubean,hydrorubeanic acid,oxaldiimidic acid, dithio,usaf mk-6
2777982
ethanedithioamide
C(=S)(C(=S)N)N
79-40-3
C2H4N2S2
120.188
OAEGRYMCJYIXQT-UHFFFAOYSA-N
dithiooxamide,rubeanic acid,ethanebis thioamide,oxamide, dithio,dithioxamide,rubeane,rubean,hydrorubeanic acid,oxaldiimidic acid, dithio,usaf mk-6
2777982
ethanedithioamide
C(=S)(C(=S)N)N
Melting Point 200.0°C
Elemental Analysis 22.7 to 23.9% (N) 19.5 to 20.5% (C)
Quantity 25g
Sulfated Ash 2% max.
Infrared Spectrum Authentic
Linear Formula H2NCSCSNH2
Solubility Information Solubility in water: 0.37g/L (20°C).
Formula Weight 120.19
Percent Purity 98%
Chemical Name or Material Dithiooxamide

GHS H Statement Harmful if swallowed. Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning