Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dithiooxamide, 98%

Click to view available options
Quantity:
5g
25g
79-40-3
C2H4N2S2
120.188
MFCD00004941
OAEGRYMCJYIXQT-UHFFFAOYSA-N
dithiooxamide,rubeanic acid,ethanebis thioamide,oxamide, dithio,dithioxamide,rubeane,rubean,hydrorubeanic acid,oxaldiimidic acid, dithio,usaf mk-6
2777982
ethanedithioamide
C(=S)(C(=S)N)N
79-40-3
C2H4N2S2
120.188
MFCD00004941
OAEGRYMCJYIXQT-UHFFFAOYSA-N
dithiooxamide,rubeanic acid,ethanebis thioamide,oxamide, dithio,dithioxamide,rubeane,rubean,hydrorubeanic acid,oxaldiimidic acid, dithio,usaf mk-6
2777982
ethanedithioamide
C(=S)(C(=S)N)N
Applications
Dithiooxamide acts as a chelating agent and used in the determination of copper(II), nickel(II) and cobalt(II). It is used as a building block in the synthesis of cyclen. It is involved in the preparation of thiazolothiazole-linked porous organic polymers and N,N′-disubstituted dithiooxamides. It is also employed as a modifier to prepare the modified glassy carbon electrode which is used to investigate the electrochemical properties of quercetin, an important flavonoid derivative. Further, it is involved in the preparation of chelating resin with formaldehyde, which finds application in separation and concentration of silver ions.

Solubility
Soluble in acetone and chloroform. Insoluble in water.

Notes
Incompatible with oxidizing agents.
Quantity 25g
Melting Point 245°C (decomposition)
Formula Weight 120.2
Percent Purity 98%
Chemical Name or Material Dithiooxamide

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : RP1575000

TSCA : TSCA