Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Dithiothreitol, >97%, Thermo Scientific™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Plastic bottle
3483-12-3
C4H10O2S2
154.24
MFCD00004877
VHJLVAABSRFDPM-IMJSIDKUSA-N
dithiothreitol,dl-1,4-dithiothreitol,dl-dithiothreitol,1,4-dithio-dl-threitol,d-1,4-dithiothreitol,d-dtt,2s,3s-1,4-dimercaptobutane-2,3-diol,threo-1,4-dimercapto-2,3-butanediol,1,4-dithiothreitol,dtt
446094
CHEBI:42170
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
C(C(C(CS)O)O)S
3483-12-3
C4H10O2S2
154.24
MFCD00004877
VHJLVAABSRFDPM-IMJSIDKUSA-N
dithiothreitol,dl-1,4-dithiothreitol,dl-dithiothreitol,1,4-dithio-dl-threitol,d-1,4-dithiothreitol,d-dtt,2s,3s-1,4-dimercaptobutane-2,3-diol,threo-1,4-dimercapto-2,3-butanediol,1,4-dithiothreitol,dtt
446094
CHEBI:42170
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
C(C(C(CS)O)O)S
Packaging Plastic bottle
Quantity 5g
Boiling Point 125°C to 130°C/12mm
Melting Point 40°C to 43°C
Flash Point >110°C (230°F)
Formula Weight 154.25
Physical Form Crystalline Powder
Percent Purity ≥97%
Chemical Name or Material Dithiothreitol