Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dithranol, BP, 98.5-101%, Spectrum™

Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
1143-38-0
C14H10O3
226.23
NUZWLKWWNNJHPT-UHFFFAOYSA-N
1,8-dihydroxy-9,10-dihydroanthracen-9-one
OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2
1143-38-0
C14H10O3
226.23
NUZWLKWWNNJHPT-UHFFFAOYSA-N
1,8-dihydroxy-9,10-dihydroanthracen-9-one
OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2

AN126, 1143-38-0, C14H10O3

Spectrum™ Dithranol, BP is used to treat chronic plaque psoriasis. Spectrum Chemical manufactured BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Loss on Drying 0.005
Sulfated Ash 0.001
Quantity 100 g
CAS Max % 1
Percent Purity 98.5 to 101%