Learn More
Tecan 50L MCA96 Tip NFil
Supplier: Tecan 30048822
Genuine Tecan 50 L disposable automation tips designed specifically for use on the Freedom EVO and Fluent MultiChannel Arm™ (MCA) 96 option. ANSI format, nested 8 stack, non-filtered. Tecan STERILE purity level - sterile, tested and certified to be free from human DNA, Dnase, Rnase, pyrogens and endotoxins. 5 tip boxes x 96 tips/wafer, 3,840 tips/sales unit. Nested disposable tips offer improved throughput and walk way time due to nested format allowing more tips per carrier. They also give you increased flexibility on the work deck due to ANSI format and are more environmentally friendly, through significant weigh and waste reduction. All MultiChannel Arm™(MCA) 96 disposable tips are designed, manufactured, tested and certified to guarantee reliable quality, helping you to acquire optimal results.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.