Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dityrosine Monoclonal Antibody (10A6), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA527566
Description
Dityrosine Monoclonal antibody specifically detects Dityrosine in Chemical samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
Dityrosine | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
-20°C | |
Liquid | |
Chemical |
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
10A6 | |
Unconjugated | |
Dityrosine; Dityrosine (DT); DT; DY | |
Synthetic Dityrosine conjugated to Keyhole Limpet Hemocyanin (KLH) | |
100 μg | |
Primary | |
Antibody | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction