Learn More
Dityrosine Mouse anti-Non-species specific, Biotin, Clone: 7D4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259360B
Catalog No. NBP259360B
Add to cart
Description
Description
Dityrosine Monoclonal antibody specifically detects Dityrosine in Non-species specific samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
Dityrosine
7D4
Western Blot, ELISA
Purified
Synthetic Dityrosine conjugated to Keyhole Limpet Hemocyanin (KLH).
Protein G purified
Store at 4C in the dark.
Monoclonal
Non-species specific
Western Blot, ELISA
Biotin
PBS with 0.05% Sodium Azide
Mouse
IgG1
100 ul
Primary
Specific for dityrosine modified proteins. Does not cross-react with 3,5-dibromotyrosine or bromotyrosine modified proteins.