Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dityrosine Mouse anti-Non-species specific, Clone: 7D4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259360

Catalog No. NBP259360


Add to cart

Description

Description

Dityrosine Monoclonal antibody specifically detects Dityrosine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Dityrosine
7D4
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50
Purified
Synthetic Dityrosine conjugated to Keyhole Limpet Hemocyanin (KLH).
Protein G purified
RUO
Primary
Specific for dityrosine modified proteins. Does not cross-react with 3,5-dibromotyrosine or bromotyrosine modified proteins.
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Unconjugated
PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide
Mouse
IgG1
100 ug
Store at -20C.
Monoclonal
Non-species specific
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only