Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dityrosine Antibody (10A6), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259361PE
Description
Dityrosine Monoclonal specifically detects Dityrosine in Non-species specific samples. It is validated for Flow Cytometry.Specifications
Dityrosine | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
Synthetic Dityrosine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
100 μL | |
Specific for dityrosine modified proteins. Does not cross-react with 3,5-dibromotyrosine or bromotyrosine modified proteins. | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
10A6 | |
Flow Cytometry | |
Mouse | |
Protein G purified | |
Primary | |
Non-species specific | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction