Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Hydroponics Growing System
SCI_ED

Self–contained ebb and flow hydroponics center with grow lights and water pump.

Supplier:  Diversified Spaces™ HGC52

Catalog No. S14215


Add to Cart

Description

Description

The perfect way to learn how plants can grow without soil and helps teach the basic concepts for the plants needs, nutrition, food production, recycling, agricultural technology and soil-less growing environments.

  • Mobile hydroponics center includes grow lights and water pump
  • Adjustable timers enable plant growth with the perfect amount of water and sunlight
  • Construction allows easy access for students and teachers alike
  • Made of solid maple and maple veneers
  • Containment plant tray is 8 x 27.5 x 42 in. (H x D x W)
  • 25 gallon nutrient reservoir with lid, a locked cabinet for timers and air pump, and a 150 watt grow light
  • Growing system is completed with a submersible pump, an air pump, an air stone, timers (2), hoses and ten 6 inch net pots
  • Unit moves easily on 4 inch locking, leveling casters
  • Overall dimensions (W x D x H): 52 x 28 x 78 in.
Specifications

Specifications

Hydroponics Growing System
Learn how plants can grow without soil; helps teach basic concepts for plants needs, nutrition, food production, recycling, agricultural technology and soil-less growing environments
Containment plant tray: 69.8D x 106.6W x 20.3cm (27.5 x 42 x 8 in.)
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit