Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Mobile Skeleton Cabinet
SCI_ED

For rolling– and hanging–style skeletons.

Manufacturer:  Diversified Spaces™ 3772422K

Catalog No. S94904MF


Add to cart

Description

Description

  • Wood-framed glass doors for easy viewing
  • Unit can be locked for security purposes
  • Shelf included for extra storage
  • Roll-in model provides extra space to accommodate a rolling stand
  • Includes retractable glide for model
  • Dimensions: 22L x 24W x 84 in.H (55.9 x 61 x 213.4cm)
Specifications

Specifications

Oak
55.8 cm
84 in.
Wood, Glass
24 in.
60.96 cm
22 in.
To store skeletons that are on a rolling stand
213.36 cm
220 lb.
99.7 kg
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit