Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Woodcrafts™ Art/Planning Table Available on GSA/VA Contract for Federal Government customers only. Products for Science Education

Designed to be used in most any environment from art and general planning to sewing and model construction. The base is constructed of two tapered solid maple pedestal style panel legs and two solid maple truss bars. The unique way the top is attached prevents the table from racking or rocking.

Manufacturer:  Diversified Woodcrafts™ PT62M

 View more versions of this product

Catalog No. S01717MF


Add to cart

Description

Description

  • 60W x 42D, with 26 and 30 in. height options
  • Constructed of solid maple with plastic laminate or maple top 
  • SEFA compliant and MAS Certified Green
  • Finished with a durable earth-friendly UV finish
  • Limited Lifetime Warranty
  • Made in the USA
  • Minimal assembly required

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Maple
42 in.
Earth-friendly UV
Any environment from art and general planning to sewing and model construction
76.2cm
152.4cm
Planning Table
60 in.
152.4cm
Northwoods
106.68cm
60 x 60 x 30 in. (152.4 x 152.4 x 76.2cm)
30 in.
60 in.
Wood Base/Maple Butcher Block Top
135 lb.
86.18kg
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit