Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diversified Spaces™ Shain Drawing Table

Units are finished with an earth-friendly UV finish.
Supplier: Diversified Spaces™ DT9SA37
Description
- Unit constructed of solid maple with 2.25 in. maple legs
Specifications
24 in. | |
Earth-friendly UV | |
37 in. | |
36 in. | |
Maple | |
24 in. | |
60.9 cm |
60.9 cm | |
36 x 24 x 37 in. (91.4 x 60.9 x 93.9 cm) | |
93.9 cm | |
91.4 cm | |
80 lb. | |
36.2 kg | |
Adjustable Drawing Surface |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title