Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Divinyl Adipate (stabilized with MEHQ) 99.0+%, TCI America™

Click to view available options
Quantity:
25g
100g
500g
4074-90-2
C10H14O4
198.218
MFCD00059202
JZQAAQZDDMEFGZ-UHFFFAOYSA-N
Adipic Acid Divinyl Ester
19997
bis(ethenyl) hexanedioate
C=COC(=O)CCCCC(=O)OC=C
4074-90-2
C10H14O4
198.218
MFCD00059202
JZQAAQZDDMEFGZ-UHFFFAOYSA-N
Adipic Acid Divinyl Ester
19997
bis(ethenyl) hexanedioate
C=COC(=O)CCCCC(=O)OC=C
Color White
Quantity 500g
Formula Weight 198.22
Physical Form Crystal-Lump at 20°C
Percent Purity ≥99.0% (GC)
Chemical Name or Material Divinyl Adipate (stabilized with MEHQ)

EINECSNumber : (2)-0870

RTECSNumber : AV1800000

TSCA : No