Alfa Aesar™ Divinyltin dichloride

Manufacturer: Alfa Aesar™ 3961309

Quantity 10g

 View more versions of this product

Catalog No. AA3961309

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7532-85-6
    Formula Weight 243.69
    MDL Number MFCD00058728
    UN Number UN2788
    Synonym Dichlorodivinyltin
    Chemical Name or Material Divinyltin dichloride

    Specifications

    Formula Weight 243.69
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00058728
    Health Hazard 2 GHS H Statement
    H300-H310-H330
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Sensitivity Moisture Sensitive
    Quantity 10g
    UN Number UN2788
    Synonym Dichlorodivinyltin
    Boiling Point 54° to 56°C (3mmHg)
    Physical Form Liquid
    CAS 7532-85-6
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
    Chemical Name or Material Divinyltin dichloride

    GHS H Statement
    H300-H310-H330
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit