Learn More
Dizocilpine maleate, 99+%, Thermo Scientific Chemicals
Description
Dizocilpine maleate is used to interact at the NMDA-associated ion channel, preventing Ca2+ flux and blocking NMDA-induced depolarizations. (+)-MK 801 demonstrates potent neuroprotective effects against excitotoxic insult, showing substantial protection against neuron loss produced by NMDA and quinolinic acid introduction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dizocilpine maleate is used to interact at the NMDA-associated ion channel, preventing Ca2+ flux and blocking NMDA-induced depolarizations. (+)-MK 801 demonstrates potent neuroprotective effects against excitotoxic insult, showing substantial protection against neuron loss produced by NMDA and quinolinic acid introduction.
Solubility
Soluble in ethanol.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Color | White |
Quantity | 10 mg |
Merck Index | 14,3386 |
Solubility Information | Soluble in ethanol. |
Formula Weight | 337.37 |
Percent Purity | ≥99% |
Physical Form | Crystalline Powder |
Chemical Name or Material | Dizocilpine maleate |
Safety and Handling
EINECSNumber | 278-614-5 |
RTECSNumber | HP1093575 |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.