dj Ortho™ Knee Sleeves

Manufacturer: Moore Medical Corp 89068

 View all options for this product

Catalog No. 19-898-850

  Check Availability
Add to Cart


    Length (English) 10 in.
    Length (Metric) 25.4cm
    Size Large
    Color Blue/black
    Design Contoured; Open patella
    Product Type dj Ortho™ Knee Sleeve
    For Use With (Application) Provides even support and warmth for controlled uniform compression
    Material Neoprene

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit