DJ Orthopedics™ Perforated Universal Knee Immobilizer
Manufacturer: Moore Medical Corp 58446
 View more versions of this product
Catalog No. 19-898-256
  Check Availability
Add to Cart
    Specifications
    Size 14 in.
    Construction Open-cell Foam with a Nylon laminated exterior, Web-Lok performance Elastic as a closure and security
    Product Type Perforated Knee Immobilizer
    For Use With (Application) Can be easily mold for a custom fit
    Includes Three posterior stays
  • Description & Specifications
Related Products
Product Content Feedback