Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Dkk-1 Antibody Pair [HRP]
Mouse Monoclonal Antibody Pair
$832.00
Specifications
Assay Range | 39.06 pg/mL |
---|---|
Assay Sensitivity | 39.06-2500 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Immunoassay Kit Format | Sandwich ELISA Kit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB010371
|
Novus Biologicals™
NBP27966615PLATES |
15 Plates |
Each of 1 for $832.00
|
|
|||||
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Dkk-1.Specifications
39.06 pg/mL | |
Sandwich ELISA | |
Sandwich ELISA Kit | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Dkk-1. | |
Sandwich ELISA | |
22943 | |
Mouse | |
Immunology, Innate Immunity, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
Dkk-1 |
39.06-2500 pg/mL | |
HRP | |
Antibody Pairs | |
Human | |
dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, DKK-1, hDkk-1, SKdickkopf-1 like | |
DKK1 | |
RUO | |
Storage is content dependent. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title