Learn More
Enzo Life SciencesSupplier Diversity Partner Dkk-1 (human), ELISA kit (1x96wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI900151
First-to-market Dkk-1 ELISA commercially available for wnt signaling pathway research. The Dkk-1 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Absorbance is read at 450 nm. The Dkk-1 (human), EIA kit offers ultra-sensitive and accurate measurement of Dickkopf-1 and has been validated in-house in normal and pathological samples. Alternative Name: Dickkopf-1. Sensitivity: 0.98 pg/ml (range 7.81-500 pg/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human. Contents: Microtiter plate, Conjugate, Antibody, Assay buffer 10 Concnetrate, Wash buffer concentrate, Standard, TMB Substrate, Stop solution 2
Catalog No. 50-658-178