Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Dkk-3 Antibody Pair [HRP]
Polyclonal/Monoclonal Antibody Pair
$1183.00
Specifications
Assay Range | 15.625 pg/mL |
---|---|
Assay Sensitivity | 15.63-1000 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Immunoassay Kit Format | Sandwich ELISA Kit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB010137
|
Novus Biologicals™
NBP27954915PLATES |
15 Plates |
Each of 1 for $1,183.00
|
|
|||||
Description
This solid phase sandwich ELISA utilizes an antibody specific for Mouse Dkk-3.Specifications
15.625 pg/mL | |
Sandwich ELISA | |
Sandwich ELISA Kit | |
This solid phase sandwich ELISA utilizes an antibody specific for Mouse Dkk-3. | |
Sandwich ELISA | |
27122 | |
Rabbit | |
RUO | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1000 pg/mL is recommended. |
15.63-1000 pg/mL | |
HRP | |
Antibody Pairs | |
Mouse | |
dickkopf (Xenopus laevis) homolog 3, dickkopf 3, dickkopf homolog 3 (Xenopus laevis), Dickkopf-3, dickkopf-related protein 3, Dkk-3, hDkk-3, regulated in glioma, REICdkk-3, RIG, RIG-like 5-6, RIG-like 7-1 | |
DKK3 | |
Signal Transduction | |
Dkk-3 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title