Learn More
Dkk-3 Mouse anti-Human, DyLight 550, Clone: 8A5C6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237430R
Catalog No. NBP237430R
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Dkk-3
8A5C6
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Purified
DKK3
Purified recombinant fragment of human DKK3 (AA: 91-350 ) expressed in E. Coli.
Protein A or G purified
Signal Transduction
Primary
27122
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
dickkopf (Xenopus laevis) homolog 3, dickkopf 3, dickkopf homolog 3 (Xenopus laevis), Dickkopf-3, dickkopf-related protein 3, Dkk-3, hDkk-3, regulated in glioma, REICdkk-3, RIG, RIG-like 5-6, RIG-like 7-1
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human