Learn More
Dkk-3 Rat anti-Mouse, Alexa Fluor™ 532, Clone: RM0221-9E12, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212265AF532
Catalog No. NP21226A32
Add to cart
Description
Description
Dkk-3 Monoclonal antibody specifically detects Dkk-3 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Dkk-3
RM0221-9E12
Western Blot
dickkopf (Xenopus laevis) homolog 3, dickkopf 3, dickkopf homolog 3 (Xenopus laevis), Dickkopf-3, dickkopf-related protein 3, Dkk-3, hDkk-3, regulated in glioma, REICdkk-3, RIG, RIG-like 5-6, RIG-like 7-1
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
DKK3
Mouse Dkk3
0.1 mL
Primary
27122