Learn More
Dkk-4 Mouse anti-Human, Alexa Fluor™ 594, Clone: 9M19, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222018AF594
Catalog No. NP222018A94
Add to cart
Description
Description
Dkk-4 Monoclonal antibody specifically detects Dkk-4 in Human samples. It is validated for Western Blot,ELISA.
Specifications
Specifications
Dkk-4
9M19
Western Blot, ELISA
dickkopf (Xenopus laevis) homolog 4, dickkopf homolog 4 (Xenopus laevis), dickkopf-4, dickkopf-related protein 4, DKK-4, hDkk-4, MGC129562, MGC129563
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
DKK4
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant Dkk-4. Mouse anti human Dkk-4 Q9UBT3.
0.1 mL
Primary
27121