Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DKK1 Monoclonal Antibody (C5)
Mouse Monoclonal Antibody
$213.50 - $522.50
Specifications
Antigen | DKK1 |
---|---|
Clone | C5 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541581
|
Invitrogen™
MA541581 |
200 μL |
Each for $522.50
|
|
|||||
PIMA543857
|
Invitrogen™
MA543857 |
20 μL |
Each for $213.50
|
|
|||||
Description
DKK1 Monoclonal Antibody for Western Blot
DKK-1 is a member of the DKK protein family which also includes DKK-2, DKK-3 and DKK-4. DKK-1 was originally identified as a Xenopus head-forming molecule that behaves as an antagonist for Wnt signaling. Subsequent studies have shown that DKK-1 and DKK-4 play important regulatory roles in the Wnt/beta-catenin signaling pathway by forming inhibitory complexes with LDL receptor-related proteins 5 and 6 (LRP5 and LRP6), which are essential components of the Wnt/beta-catenin signaling system. LRP5 and LRP6 are single-pass transmembrane proteins that appear to act as co-receptors for Wnt ligands involved in the Wnt/beta-catenin signaling cascade. It has been suggested that by inhibiting Wnt/beta-catenin signaling, which is essential for posterior patterning in vertebrates, DKK-1 permits anterior development. This notion is supported by the finding that mice deficient of DKK-1 expression lack head formation and die during embryogenesis.Specifications
DKK1 | |
1 mg/mL | |
Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
O94907 | |
22943 | |
Recombinant protein Dickkopf Related Protein 1. The antigen corresponds to amino acid range 33-266 of the target protein. | |
Antibody |
C5 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2b κ | |
Human | |
DKK1 | |
dickkopf 1 homolog; dickkopf homolog 1; dickkopf homolog 1 (Xenopus laevis); dickkopf related protein-1; dickkopf WNT signaling pathway inhibitor 1; Dickkopf1; dickkopf-1; dickkopf-1 like; dickkopf-like protein 1; dickkopf-related protein 1; DKK1; DKK-1; hDkk1; hDkk-1; mdkk-1; SK; UNQ492/PRO1008 | |
DKK1 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title