Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DKKL1 Polyclonal Antibody
DKKL1 Polyclonal Antibody
Supplier: Thermo Scientific PA561463
Description
DKKL1 Polyclonal Antibody for ICC/IF, IHC

Specifications
DKKL1 | |
Polyclonal | |
Unconjugated | |
DKKL1 | |
AI414495; Cancer/testis antigen 34; CT34; dickkopf like acrosomal protein 1; dickkopf-like 1; dickkopf-like 1 (soggy); dickkopf-like protein 1; Dkkl1; mSgy; Protein soggy-1; RGD1563523; SGY; SGY1; SGY-1; Soggy; soggy 1; soggy-1; testicular secretory protein Li 15; UNQ735/PRO1429 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27120 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UK85 | |
DKKL1 | |
Recombinant protein corresponding to Human DKKL1. Recombinant protein control fragment (Product #RP-90048). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction