Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2,3-Diaminopropionic acid monohydrochloride, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
54897-59-5
C3H8N2O2·HCl
140.57
MFCD00012884
SKWCZPYWFRTSDD-UHFFFAOYSA-N
2,3-diaminopropanoic acid hydrochloride, dl-2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine hydrochloride, dl-2,3-diaminopropionic acid monohydrochloride, 2,3-diaminopropionic acid hydrochloride, +--2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine monohydrochloride, 2,3-diaminopropionic acid monohydrochloride, dl-2,3-diaminopropanoic acid hydrochloride, diaminopropionic acid
108638
2,3-diaminopropanoic acid;hydrochloride
C(C(C(=O)O)N)N.Cl
54897-59-5
C3H8N2O2·HCl
140.57
MFCD00012884
SKWCZPYWFRTSDD-UHFFFAOYSA-N
2,3-diaminopropanoic acid hydrochloride, dl-2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine hydrochloride, dl-2,3-diaminopropionic acid monohydrochloride, 2,3-diaminopropionic acid hydrochloride, +--2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine monohydrochloride, 2,3-diaminopropionic acid monohydrochloride, dl-2,3-diaminopropanoic acid hydrochloride, diaminopropionic acid
108638
2,3-diaminopropanoic acid;hydrochloride
C(C(C(=O)O)N)N.Cl
Melting Point 222°C to 226°C
Packaging Glass bottle
Color White to Beige
Quantity 25g
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula H2NCH2CH(NH2)CO2H·HCl
Beilstein 04, 405
Merck Index 10, 2948
Solubility Information Solubility in water: freely soluble in water
Formula Weight 140.57
Physical Form Fine Crystalline Powder
Percent Purity 99%
Chemical Name or Material DL-2, 3-Diaminopropionic acid monohydrochloride
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 259-387-1