Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2,3-Dimercapto-1-propanesulfonic acid, sodium salt monohydrate, 95%, ACROS Organics™

Click to view available options
Quantity:
250mg
Packaging:
Glass bottle
207233-91-8
C3H9NaO4S3
228.271
MFCD00149543
XMUHNMQFDVIWGU-UHFFFAOYSA-M
sodium 2,3-dimercaptopropanesulfonate monohydrate,unithiol,dl-2,3-dimercapto-1-propanesulfonic acid sodium salt monohydrate,sodium 2,3-dimercaptopropane-1-sulfonate hydrate,2,3-dimercapto-1-propanesulfonic acid sodium salt monohydrate,sodium 2,3-dimercaptopropanesulfonate hydrate,dmps,sodium 2,3-disulfanylpropane-1-sulfonate hydrate,unitiol monohydrate
23676755
sodium;2,3-bis(sulfanyl)propane-1-sulfonate;hydrate
C(C(CS(=O)(=O)[O-])S)S.O.[Na+]
207233-91-8
C3H9NaO4S3
228.271
MFCD00149543
XMUHNMQFDVIWGU-UHFFFAOYSA-M
sodium 2,3-dimercaptopropanesulfonate monohydrate,unithiol,dl-2,3-dimercapto-1-propanesulfonic acid sodium salt monohydrate,sodium 2,3-dimercaptopropane-1-sulfonate hydrate,2,3-dimercapto-1-propanesulfonic acid sodium salt monohydrate,sodium 2,3-dimercaptopropanesulfonate hydrate,dmps,sodium 2,3-disulfanylpropane-1-sulfonate hydrate,unitiol monohydrate
23676755
sodium;2,3-bis(sulfanyl)propane-1-sulfonate;hydrate
C(C(CS(=O)(=O)[O-])S)S.O.[Na+]
Infrared Spectrum Authentic
Color White
Loss on Drying 7% to 9% (1 g, 100°C)
Quantity 250mg
Packaging Glass bottle
Assay Percent Range 95%
Linear Formula HSCH2CH(SH)CH2SO3Na·H2O
Merck Index 14, 3210
Formula Weight 228.28
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material DL-2, 3-Dimercapto-1-propanesulfonic acid, sodium salt monohydrate, 95%
Show More Show Less

AIR SENSITIVE

EINECSNumber : 223-796-3

RTECSNumber : TZ6420000