Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Amino-1-propanol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
6168-72-5
C3H9NO
75.111
MFCD00008085
BKMMTJMQCTUHRP-UHFFFAOYSA-N
dl-alaninol,2-aminopropanol,2-amino-1-propanol,dl-2-amino-1-propanol,1-propanol, 2-amino,1-hydroxy-2-aminopropane,2-amino-2-methylethanol,alaninol,propanol, 2-amino,1-methyl-2-hydroxyethylamine
5126
2-aminopropan-1-ol
CC(CO)N
6168-72-5
C3H9NO
75.111
MFCD00008085
BKMMTJMQCTUHRP-UHFFFAOYSA-N
dl-alaninol,2-aminopropanol,2-amino-1-propanol,dl-2-amino-1-propanol,1-propanol, 2-amino,1-hydroxy-2-aminopropane,2-amino-2-methylethanol,alaninol,propanol, 2-amino,1-methyl-2-hydroxyethylamine
5126
2-aminopropan-1-ol
CC(CO)N
Refractive Index 1.4480 to 1.4510
Specific Rotation -0.5° to +0.5° (20°C, 589nm) (neat)
Infrared Spectrum Authentic
Packaging Glass bottle
Flash Point 83°C
Color Colorless to Yellow
Boiling Point 173.0°C to 176.0°C
Density 0.9430g/mL
Quantity 5g
Melting Point 8.0°C
Assay Percent Range 98%
Linear Formula CH3CH(NH2)CH2OH
Beilstein 04, I, 432
Merck Index 10, 472
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in alcohol and ether
Formula Weight 75.11
Physical Form Viscous Liquid
Percent Purity 98%
Chemical Name or Material DL-2-Amino-1-propanol, 98%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 228-207-3

TSCA : TSCA