Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Amino-3-methyl-1-butanol, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
16369-05-4
C5H13NO
103.16
MFCD00004730
NWYYWIJOWOLJNR-UHFFFAOYSA-N
2-amino-3-methyl-1-butanol,dl-valinol,valinol,l-+-valinol,2-amino-3-methyl-butan-1-ol,1-butanol, 2-amino-3-methyl,1-butanol, 2-amino-3-methyl-, .+/-.,s-+-2-amino-3-methyl-1-butanol,1-2-amino-3-methylbutan-1-ol
79019
2-amino-3-methylbutan-1-ol
CC(C)C(CO)N
16369-05-4
C5H13NO
103.16
MFCD00004730
NWYYWIJOWOLJNR-UHFFFAOYSA-N
2-amino-3-methyl-1-butanol,dl-valinol,valinol,l-+-valinol,2-amino-3-methyl-butan-1-ol,1-butanol, 2-amino-3-methyl,1-butanol, 2-amino-3-methyl-, .+/-.,s-+-2-amino-3-methyl-1-butanol,1-2-amino-3-methylbutan-1-ol
79019
2-amino-3-methylbutan-1-ol
CC(C)C(CO)N
Specific Gravity 0.93
Infrared Spectrum Authentic
Flash Point 90°C
Packaging Glass bottle
Color Yellow
Boiling Point 75.0°C to 77.0°C (8.0mmHg)
Refractive Index 1.4533 to 1.4553
Quantity 5g
Density 0.9300g/mL
Assay Percent Range 96% min. (GC)
Linear Formula (CH3)2CHCH(NH2)CH2OH
Fieser 12,563; 13,341; 01,63; 02,32; 03,29; 04,41; 05,48; 09,59; 11,7
Formula Weight 103.16
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material DL-2-Amino-3-methyl-1-butanol, 95%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 240-425-0