Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dl-2-amino-3-phenyl-1-propanol, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
16088-07-6
C9H13NO
151.21
MFCD00066689
STVVMTBJNDTZBF-UHFFFAOYSA-N
dl-phenylalaninol,2-amino-3-phenyl-1-propanol,phenylalaninol,s-phenylalaninol,l-2-amino-3-phenyl-1-propanol,s-2-benzylethanolamine,benzenepropanol, beta-amino,s-beta-aminobenzenepropanol,chembl21676,2-amino-3-phenyl-propan-1-ol
76652
2-amino-3-phenylpropan-1-ol
C1=CC=C(C=C1)CC(CO)N
16088-07-6
C9H13NO
151.21
MFCD00066689
STVVMTBJNDTZBF-UHFFFAOYSA-N
dl-phenylalaninol,2-amino-3-phenyl-1-propanol,phenylalaninol,s-phenylalaninol,l-2-amino-3-phenyl-1-propanol,s-2-benzylethanolamine,benzenepropanol, beta-amino,s-beta-aminobenzenepropanol,chembl21676,2-amino-3-phenyl-propan-1-ol
76652
2-amino-3-phenylpropan-1-ol
C1=CC=C(C=C1)CC(CO)N
Quantity 1g
Melting Point 64.0°C to 72.0°C
Color White to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula C6H5CH2CH(NH2)CH2OH
Beilstein 13, III, 1757
Solubility Information Solubility in water: soluble. Other solubilities: soluble in diethyl ether, benzene, petroleum ether
Formula Weight 151.21
Physical Form Crystalline Powder and Chunks
Percent Purity 98%
Chemical Name or Material DL-2-Amino-3-phenyl-1-propanol
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger