Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Aminobutyric acid, 99%, Acros Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
2835-81-6
C4H9NO2
103.12
QWCKQJZIFLGMSD-UHFFFAOYSA-N
dl-2-aminobutyric acid,2-aminobutyric acid,alpha-aminobutyric acid,butanoic acid, 2-amino,butyrine,dl-2-amino-n-butyric acid,h-dl-abu-oh,aaba,dl-alpha-amino-n-butyric acid,alpha-amino-n-butyric acid
6657
CHEBI:35621
2-aminobutanoic acid
CCC(C(=O)O)N
2835-81-6
C4H9NO2
103.12
QWCKQJZIFLGMSD-UHFFFAOYSA-N
dl-2-aminobutyric acid,2-aminobutyric acid,alpha-aminobutyric acid,butanoic acid, 2-amino,butyrine,dl-2-amino-n-butyric acid,h-dl-abu-oh,aaba,dl-alpha-amino-n-butyric acid,alpha-amino-n-butyric acid
6657
CHEBI:35621
2-aminobutanoic acid
CCC(C(=O)O)N
Color White
Melting Point 291.0°C to 293.0°C
Packaging Glass bottle
Quantity 25g
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula C2H5CH(NH2)CO2H
Beilstein 04,408
Merck Index 15,423
Solubility Information Solubility in water: 210g/L (25 c). Other solubilities: sparingly soluble in alcohol,insoluble in ether
Formula Weight 103.12
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material DL-2-Aminobutyric acid, 99+%
Show More Show Less

EINECSNumber : 220-616-5