Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Isopropylserine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
7522-43-2
C6H13NO3
147.17
MFCD01863303
QTYSAGIKKRDGIV-ZCFIWIBFSA-N
dl-2-isopropylserine,s-2-amino-2-hydroxymethyl-3-methylbutanoic acid,2-isopropyl-l-serine,2s-2-amino-2-hydroxymethyl-3-methylbutanoic acid
7010084
(2S)-2-amino-2-(hydroxymethyl)-3-methylbutanoic acid
CC(C)C(CO)(C(=O)O)N
7522-43-2
C6H13NO3
147.17
MFCD01863303
QTYSAGIKKRDGIV-ZCFIWIBFSA-N
dl-2-isopropylserine,s-2-amino-2-hydroxymethyl-3-methylbutanoic acid,2-isopropyl-l-serine,2s-2-amino-2-hydroxymethyl-3-methylbutanoic acid
7010084
(2S)-2-amino-2-(hydroxymethyl)-3-methylbutanoic acid
CC(C)C(CO)(C(=O)O)N
Melting Point 262°C
Infrared Spectrum Authentic
Color Cream
Packaging Glass bottle
Quantity 5g
Assay Percent Range 99%
Solubility Information Solubility in water: .. Other solubilities: soluble in aq. acids and bases
Formula Weight 147.17
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material DL-2-Isopropylserine, 99%
Show More Show Less