Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Methoxy-alpha-methylbenzyl alcohol, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
13513-82-1
C9H12O2
152.193
MFCD00004512
DHHGVIOVURMJEA-UHFFFAOYSA-N
1-2-methoxyphenyl ethanol,1-2-methoxyphenyl ethan-1-ol,1-o-methoxyphenyl ethanol,1-2-methoxy-phenyl-ethanol,2-methoxy-alpha-methylbenzyl alcohol,dl-o-methoxy-alpha-methylbenzylalcohol,o-methoxy-.alpha.-methylbenzyl alcohol,o-methoxyphenyl ethanol,acmc-1bdow,1 o-methoxyphenyl ethanol
114538
1-(2-methoxyphenyl)ethanol
CC(C1=CC=CC=C1OC)O
13513-82-1
C9H12O2
152.193
MFCD00004512
DHHGVIOVURMJEA-UHFFFAOYSA-N
1-2-methoxyphenyl ethanol,1-2-methoxyphenyl ethan-1-ol,1-o-methoxyphenyl ethanol,1-2-methoxy-phenyl-ethanol,2-methoxy-alpha-methylbenzyl alcohol,dl-o-methoxy-alpha-methylbenzylalcohol,o-methoxy-.alpha.-methylbenzyl alcohol,o-methoxyphenyl ethanol,acmc-1bdow,1 o-methoxyphenyl ethanol
114538
1-(2-methoxyphenyl)ethanol
CC(C1=CC=CC=C1OC)O
Quantity 10g
Infrared Spectrum Authentic
Color White
Density 1.0800g/mL
Packaging Glass bottle
Melting Point 37°C to 39°C
Assay Percent Range 99%
Linear Formula CH3OC6H4CH(CH3)OH
Beilstein 06, 903
Formula Weight 152.19
Physical Form Crystalline Mass
Percent Purity ≥98.5%
Chemical Name or Material DL-2-Methoxy-α-methylbenzyl alcohol, 99%
Show More Show Less

EINECSNumber : 236-850-6